Search Thermo Fisher Scientific
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147153.3 | NP_671495.1 | 12 | 1252 | ||
NM_178851.3 | NP_849182.1 | 11 | 1219 | |||
GenBank mRNA | AB028844.1 | - | 11 | 1199 | ||
AF281050.1 | - | 12 | 1229 | |||
AF281051.1 | - | 11 | 1196 | |||
AK038824.1 | - | 11 | 1208 | |||
AK042253.1 | - | 11 | 1208 | |||
AK084743.1 | - | 11 | 1219 | |||
AK129210.1 | - | 11 | 1214 | |||
AK140041.1 | - | 12 | 1257 | |||
AK141424.1 | - | 11 | 1202 | |||
AK154103.1 | - | 11 | 1210 | |||
AK159889.1 | - | 11 | 1212 | |||
AK160109.1 | - | 11 | 1212 | |||
AK170831.1 | - | 12 | 1279 | |||
AK171008.1 | - | 11 | 1208 | |||
BC007479.1 | - | 11 | 1170 | 3492875 |