Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Map3k15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163085.2 | NP_001156557.2 | 15 | 2026 | ||
GenBank mRNA | BC031147.1 | - | 14 | 1531 | ||
BC043943.1 | - | 14 | 1531 |