Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173037.1 | NP_766625.1 | 13 | 2395 | ||
GenBank mRNA | AK047206.1 | - | 8 | 1113 | ||
AK054283.1 | - | 13 | 2349 | |||
AK077757.1 | - | 13 | 2380 | |||
AK085030.1 | - | 13 | 2395 | |||
AK173250.1 | - | 13 | 2302 | |||
BC066134.1 | - | 13 | 2325 | |||
BC066869.1 | - | 13 | 2325 | 30106406 | ||
BC069838.1 | - | 13 | 2325 | 30106422 |