Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTK7 SIRNA Assays ›
          Gene Symbol:
          PTK7
          Gene Name:
          protein tyrosine kinase 7 (inactive)
          Gene Aliases:
          CCK-4, CCK4
          Chromosome Location:
          Chr.6: 43076268 - 43161720 on Build GRCh38
          UniGene ID:
          Hs.90572
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11491
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTK7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270398.1 NP_001257327.1 17 2781
          NM_002821.4 NP_002812.2 17 2908
          NM_152880.3 NP_690619.1 16 2788
          NM_152881.3 NP_690620.1 14 2518
          NM_152882.3 NP_690621.1 16 2740
          NR_072997.1 16 2712
          NR_072998.1 16 2675
          XM_011514765.2 XP_011513067.1 16 2656
          XM_011514766.2 XP_011513068.1 14 2386
          GenBank mRNA AB209350.1 - 16 3924
          AF531868.1 - 17 2834
          AF531869.1 - 16 2714
          AF531870.1 - 14 2444
          AF531871.1 - 16 2666
          AF531872.1 - 16 2601
          AK055648.1 - 4 1034
          AK291016.1 - 16 2620
          AK292589.1 - 17 2835
          AL157486.1 - 5 571
          BC071557.1 - 17 2862 30347652
          BQ881986.1 - 5 709
          U33635.1 - 17 2879
          U40271.2 - 17 2834

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00177173_m1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of planar polarity
          ventricular septum development
          axis elongation
          protein phosphorylation
          cell adhesion
          signal transduction
          positive regulation of neuron projection development
          cell migration
          actin cytoskeleton reorganization
          wound healing
          establishment of epithelial cell apical/basal polarity
          convergent extension
          canonical Wnt signaling pathway
          lung-associated mesenchyme development
          coronary vasculature development
          cellular response to retinoic acid
          cochlea morphogenesis
          planar cell polarity pathway involved in neural tube closure
          protein kinase activity
          protein binding
          ATP binding
          coreceptor activity involved in Wnt signaling pathway, planar cell polarity pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0