Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN3 SIRNA Assays ›
Gene Symbol:
PTPN3
Gene Name:
protein tyrosine phosphatase, non-receptor type 3
Gene Aliases:
PTP-H1, PTPH1
Chromosome Location:
Chr.9: 109375694 - 109538395 on Build GRCh38
UniGene ID:
Hs.436429
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11513
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145368.1
NP_001138840.1
13
1196
NM_001145369.1
NP_001138841.1
9
3380
NM_001145370.1
NP_001138842.1
8
3245
NM_001145371.1
NP_001138843.1
4
814
NM_001145372.1
NP_001138844.1
3
679
NM_002829.3
NP_002820.3
14
1331
XM_006717197.3
XP_006717260.1
14
1440
XM_006717199.3
XP_006717262.1
14
1465
XM_006717201.3
XP_006717264.1
13
1272
XM_006717202.3
XP_006717265.1
11
1034
XM_006717203.3
XP_006717266.1
11
993
XM_006717204.3
XP_006717267.1
14
1367
XM_011518888.2
XP_011517190.1
14
1401
XM_011518889.2
XP_011517191.1
14
2071
XM_017014955.1
XP_016870444.1
14
1423
XM_017014956.1
XP_016870445.1
12
1144
XM_017014957.1
XP_016870446.1
13
1232
GenBank mRNA
AK295875.1
-
12
1116
AK296265.1
-
3
463
AK296388.1
-
4
740
AK304434.1
-
13
1196
AK316066.1
-
13
1196
AK316118.1
-
3
605
BC126117.1
-
13
1241
8991833
BC143848.1
-
12
1106
9052363
BX648253.1
-
8
3246
BX648735.1
-
9
1815
CR749204.1
-
2
258
DQ104439.1
-
13
1219
M64572.1
-
13
1242
S39392.1
-
6
642
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of mitotic cell cycle
negative regulation of membrane protein ectodomain proteolysis
liver regeneration
regulation of membrane depolarization during action potential
regulation of sodium ion transmembrane transporter activity
phosphotyrosine binding
protein tyrosine phosphatase activity
protein binding
cytoskeletal protein binding
sodium channel regulator activity
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0