Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081264.1 | NP_001074733.1 | 3 | 304 | ||
GenBank mRNA | AK036792.1 | - | 3 | 266 | ||
AK087075.1 | - | 3 | 271 | |||
AK087630.1 | - | 3 | 263 | |||
AK157903.1 | - | 3 | 266 | |||
AK171887.1 | - | 3 | 257 | |||
BC050854.1 | - | 3 | 269 | 6332345 | ||
BC080867.1 | - | 3 | 244 |