Search
Search

Dtd2| Gene Symbol: Dtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029545.2 | NP_083821.1 | 3 | 482 | ||
| GenBank mRNA | AK018311.1 | - | 3 | 481 | ||
| AK032228.1 | - | 3 | 480 | |||
| AK042771.1 | - | 3 | 483 | |||
| BC089589.1 | - | 3 | 482 | 6760937 | ||