Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EP300 SIRNA Assays ›
          Gene Symbol:
          Ep300
          Gene Name:
          E1A binding protein p300
          Gene Aliases:
          A430090G16, A730011L11, KAT3B, p300, p300 HAT
          Chromosome Location:
          Chr.15: 81586214 - 81652077 on Build GRCm38
          UniGene ID:
          Mm.258397
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s116227

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ep300
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_177821.6 NP_808489.4 27 4839
          GenBank mRNA AK040384.1 - 6 890
          BC144976.1 - 27 4693 9053524
          BC150681.1 - 27 4693 9087592
          BU701221.1 - 1 47
          CD348567.1 - 5 632

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to hypoxia
          somitogenesis
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein acetylation
          internal protein amino acid acetylation
          apoptotic process
          positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response
          cell cycle
          heart development
          skeletal muscle tissue development
          circadian rhythm
          response to glucose
          organ morphogenesis
          positive regulation of glycoprotein biosynthetic process
          positive regulation of gene expression
          positive regulation of cell death
          positive regulation of muscle atrophy
          histone acetylation
          N-terminal peptidyl-lysine acetylation
          internal peptidyl-lysine acetylation
          cell differentiation
          positive regulation of cell growth
          lung development
          negative regulation of cellular metabolic process
          positive regulation of cellular metabolic process
          positive regulation of protein binding
          positive regulation of collagen biosynthetic process
          positive regulation of protein import into nucleus, translocation
          cellular response to UV
          positive regulation of histone acetylation
          response to drug
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of DNA binding
          protein kinase B signaling
          response to estrogen
          positive regulation by host of viral transcription
          histone H3 acetylation
          histone H4 acetylation
          histone H2B acetylation
          positive regulation of translation
          positive regulation of axon extension
          positive regulation of cell size
          positive regulation of proteolysis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          positive regulation of protein secretion
          protein stabilization
          positive regulation of sequence-specific DNA binding transcription factor activity
          cartilage development
          response to calcium ion
          regulation of angiotensin metabolic process
          positive regulation of sarcomere organization
          negative regulation of cell death
          regulation of androgen receptor signaling pathway
          protein-DNA complex assembly
          response to dexamethasone
          regulation of tubulin deacetylation
          positive regulation of protein acetylation
          regulation of nucleic acid-templated transcription
          negative regulation of miRNA metabolic process
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          core promoter binding
          RNA polymerase II transcription factor binding
          RNA polymerase II activating transcription factor binding
          core promoter proximal region DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          p53 binding
          DNA binding
          chromatin binding
          damaged DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription cofactor activity
          transcription coactivator activity
          antigen binding
          histone acetyltransferase activity
          lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          protein binding
          beta-catenin binding
          protein C-terminus binding
          transcription factor binding
          zinc ion binding
          acetyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          protein kinase binding
          chromatin DNA binding
          protein complex binding
          activating transcription factor binding
          nuclear hormone receptor binding
          glucocorticoid receptor binding
          peroxisome proliferator activated receptor binding
          bHLH transcription factor binding
          SMAD binding
          metal ion binding
          androgen receptor binding
          mitogen-activated protein kinase binding
          NF-kappaB binding
          pre-mRNA intronic binding
          protein antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0