Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGER SIRNA Assays ›
          Gene Symbol:
          AGER
          Gene Name:
          advanced glycosylation end product-specific receptor
          Gene Aliases:
          RAGE, SCARJ1
          Chromosome Location:
          Chr.6: 32180968 - 32185247 on Build GRCh38
          UniGene ID:
          Hs.534342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1168

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGER
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136.4 NP_001127.1 5, 6 600
          NM_001206929.1 NP_001193858.1 5, 6 648
          NM_001206932.1 NP_001193861.1 5, 6 558
          NM_001206934.1 NP_001193863.1 5, 6 648
          NM_001206936.1 NP_001193865.1 5, 6 600
          NM_001206940.1 NP_001193869.1 5, 6 600
          NM_001206954.1 NP_001193883.1 5, 6 600
          NM_001206966.1 NP_001193895.1 5, 6 600
          NM_172197.2 NP_751947.1 5, 6 558
          NR_038190.1 4, 5 783
          XM_017010328.1 XP_016865817.1 5, 6 1573
          GenBank mRNA AB036432.1 - 5, 6 524
          AB061668.1 - 5, 6 524
          AB061669.1 - 4, 5 707
          AJ133822.1 - 5, 6 473
          AK298100.1 - 5, 6 602
          AK313178.1 - 5, 6 528
          AY755619.1 - 5, 6 510
          AY755620.1 - 5, 6 510
          AY755621.1 - 5, 6 558
          AY755622.1 - 5, 6 558
          AY755623.1 - 5, 6 510
          AY755624.1 - 5, 6 468
          AY755628.1 - 5, 6 510
          BC020669.1 - 5, 6 519 4718076
          DQ104251.1 - 5, 6 510
          DQ104252.1 - 5, 6 468
          DQ104253.1 - 5, 6 510
          DQ104254.1 - 4, 5 693
          EU117141.1 - 5, 6 510
          EU428812.1 - 4, 5 683
          EU428813.1 - 4, 5 683
          EU428815.1 - 4, 5 683
          EU428816.1 - 4, 5 683
          EU826618.1 - 5, 6 500
          EU826619.1 - 5, 6 548
          FJ545655.1 - 5, 6 572
          GU139370.1 - 2, 3 958
          GU139371.1 - 2, 3 957
          GU139372.1 - 4, 5 757
          GU139373.1 - 3, 4 888
          GU139374.1 - 3, 4 834
          GU139375.1 - 4, 5 757
          GU139377.1 - 4, 5 706
          GU139378.1 - 5, 6 522
          HQ148133.1 - 5, 6 524
          HQ148135.1 - 4, 5 704
          KC692917.1 - 5, 6 500
          M91211.1 - 5, 6 500

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of T cell mediated cytotoxicity
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          negative regulation of endothelial cell proliferation
          inflammatory response
          negative regulation of cell adhesion
          cell surface receptor signaling pathway
          JAK-STAT cascade
          brain development
          glycoprotein metabolic process
          response to wounding
          response to fructose
          positive regulation of autophagy
          negative regulation of endothelial cell migration
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of fibroblast migration
          response to activity
          positive regulation of smooth muscle cell migration
          lung development
          neuron projection development
          negative regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          negative regulation of collagen biosynthetic process
          response to vitamin A
          response to genistein
          negative regulation of osteoblast proliferation
          cellular response to drug
          positive regulation of activated T cell proliferation
          positive regulation of neuron apoptotic process
          innate immune response
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of NF-kappaB transcription factor activity
          response to methylglyoxal
          calcium ion homeostasis
          response to hyperoxia
          positive regulation of phagocytosis, engulfment
          transdifferentiation
          cellular response to hydrogen peroxide
          cellular response to glucose stimulus
          cellular response to fatty acid
          cellular response to organic cyclic compound
          response to selenite ion
          positive regulation of potassium ion transmembrane transporter activity
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          regulation of CD4-positive, alpha-beta T cell activation
          positive regulation of type B pancreatic cell apoptotic process
          positive regulation of dendritic cell differentiation
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          identical protein binding
          S100 protein binding
          advanced glycation end-product receptor activity
          high mobility group box 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0