Search
Search

Serpina7| Gene Symbol: Serpina7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177920.5 | NP_808588.3 | 5 | 1230 | ||
| GenBank mRNA | AK050361.1 | - | 6 | 775 | ||
| AK149459.1 | - | 5 | 1211 | |||
| AY575783.1 | - | 4 | 1164 | |||
| BC132100.1 | - | 4 | 1225 | 40130377 | ||