Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037906.2 | NP_001032995.1 | 4 | 417 | ||
GenBank mRNA | AY622226.1 | - | 4 | 417 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK035921.1 | - | 906 | Chr. - - [-] on Build |