Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177387.3 | NP_796361.2 | 7 | 1310 | ||
GenBank mRNA | AK029782.1 | - | 6 | 701 | ||
AK086181.1 | - | 7 | 1257 | |||
AK147850.1 | - | 7 | 1312 | |||
AK162950.1 | - | 7 | 883 | |||
BC138155.1 | - | 7 | 1050 | 8861174 |