Search
Search

Agap1| Gene Symbol: Agap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037136.1 | NP_001032213.1 | 8 | 1446 | ||
| NM_178119.3 | NP_835220.1 | 8 | 1446 | |||
| GenBank mRNA | AK046523.1 | - | 8 | 1407 | ||
| AK129289.2 | - | 8 | 1393 | |||
| AK147503.1 | - | 8 | 1452 | |||
| AK147521.1 | - | 9 | 2097 | |||
| BC096588.1 | - | 8 | 1394 | |||