Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mrpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172252.3 | NP_758456.2 | 5, 6 | 494 | ||
GenBank mRNA | AK003285.1 | - | 5, 6 | 481 | ||
AK012784.1 | - | 5, 6 | 486 | |||
AK020739.1 | - | 5, 6 | 495 | |||
BC028768.1 | - | 5, 6 | 469 | 1282313 |