Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Myo18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291212.1 | NP_001278141.1 | 37 | 5601 | ||
NM_001291213.1 | NP_001278142.1 | 37 | 5709 | |||
NM_001291214.1 | NP_001278143.1 | 35 | 4556 | |||
NM_001291215.1 | NP_001278144.1 | 38 | 4622 | |||
NM_011586.3 | NP_035716.1 | 36 | 5565 | |||
GenBank mRNA | AB026497.1 | - | 36 | 5535 | ||
AK147584.1 | - | 38 | 4571 | |||
AK171342.1 | - | 35 | 4553 | |||
BC023378.1 | - | 12 | 1430 | |||
BC046638.1 | - | 19 | 2393 | |||
BC057920.1 | - | 7 | 776 | |||
BC138366.1 | - | 37 | 5543 | 8861386 | ||
BC145257.1 | - | 37 | 5651 |