Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JAG1 SIRNA Assays ›
          Gene Symbol:
          JAG1
          Gene Name:
          jagged 1
          Gene Aliases:
          AGS, AHD, AWS, CD339, HJ1, JAGL1
          Chromosome Location:
          Chr.20: 10637684 - 10674046 on Build GRCh38
          UniGene ID:
          Hs.224012
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1176

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JAG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000214.2 NP_000205.1 19, 20 2886
          XM_017027707.1 XP_016883196.1 18, 19 2991
          GenBank mRNA AF003837.1 - 19, 20 2829
          AF028593.1 - 19, 20 2633
          AK302554.1 - 17, 18 2236
          AK312722.1 - 19, 20 2455
          BC098393.1 - 19, 20 2789 30528888
          BC126205.1 - 20, 21 2394 8991921
          BC126207.1 - 19, 20 2622 8991923
          U61276.1 - 19, 20 2740
          U73936.1 - 19, 20 2783
          U77720.1 - 19, 20 2332
          U77914.1 - 2, 3 144

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell fate determination
          blood vessel remodeling
          morphogenesis of an epithelial sheet
          T cell mediated immunity
          pulmonary valve morphogenesis
          cardiac right ventricle morphogenesis
          Notch signaling pathway
          Notch receptor processing
          nervous system development
          hemopoiesis
          keratinocyte differentiation
          regulation of cell migration
          response to muramyl dipeptide
          aorta morphogenesis
          regulation of cell proliferation
          auditory receptor cell differentiation
          myoblast differentiation
          endothelial cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of myeloid cell differentiation
          negative regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          cardiac septum morphogenesis
          ciliary body morphogenesis
          pulmonary artery morphogenesis
          cardiac neural crest cell development involved in outflow tract morphogenesis
          Notch signaling involved in heart development
          endocardial cushion cell development
          nephron development
          glomerular visceral epithelial cell development
          distal tubule development
          loop of Henle development
          neuronal stem cell population maintenance
          negative regulation of stem cell differentiation
          Notch binding
          structural molecule activity
          calcium ion binding
          protein binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0