Search
Search

RANBP1| Gene Symbol: RANBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278639.1 | NP_001265568.1 | 5 | 959 | ||
| NM_001278640.1 | NP_001265569.1 | 5 | 707 | |||
| NM_001278641.1 | NP_001265570.1 | 5 | 827 | |||
| NM_002882.3 | NP_002873.1 | 5 | 707 | |||
| XM_011530290.2 | XP_011528592.1 | 5 | 1019 | |||
| XM_011530291.2 | XP_011528593.1 | 5 | 1021 | |||
| XM_017028890.1 | XP_016884379.1 | 5 | 729 | |||
| XM_017028891.1 | XP_016884380.1 | 5 | 822 | |||
| XM_017028892.1 | XP_016884381.1 | 5 | 613 | |||
| GenBank mRNA | AK094410.1 | - | 2 | 1340 | ||
| AK097659.1 | - | 6 | 1042 | |||
| AK223506.1 | - | 5 | 626 | |||
| BG717186.1 | - | 5 | 576 | |||
| CA489474.1 | - | 5 | 649 | |||
| CR456556.1 | - | 5 | 535 | |||
| D38076.1 | - | 5 | 630 | |||
| X83617.1 | - | 5 | 583 | |||