Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RASGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006909.2 | NP_008840.1 | 4 | 648 | ||
XM_005248565.1 | XP_005248622.1 | 4 | 665 | |||
XM_017009682.1 | XP_016865171.1 | 4 | 330 | |||
XM_017009683.1 | XP_016865172.1 | 4 | 665 | |||
GenBank mRNA | AF023130.1 | - | 4 | 598 | ||
AF181250.1 | - | 4 | 598 | |||
AK094809.1 | - | 1 | 523 | |||
BC126112.1 | - | 4 | 648 | 8991828 | ||
BC136296.1 | - | 4 | 648 | 9020283 | ||
BC143870.1 | - | 4 | 648 |