Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207544.1 | NP_997427.1 | 1 | 417 |
Gene Symbol: Vmn1r186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167567.1 | NP_001161039.1 | 1 | 387 |
Gene Symbol: Vmn1r60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166732.1 | NP_001160204.1 | 1 | 387 |
Gene Symbol: Vmn1r62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030741.2 | NP_109666.2 | 1 | 1105 | ||
GenBank mRNA | BC113757.1 | - | 1 | 1082 | 40047296 | |
BC147681.1 | - | 1 | 466 | 9056654 | ||
BC147684.1 | - | 1 | 466 | 9056657 |