Search
Search

RCN2| Gene Symbol: RCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271837.1 | NP_001258766.1 | 7 | 1047 | ||
| NM_002902.2 | NP_002893.1 | 6 | 993 | |||
| GenBank mRNA | AK126522.1 | - | 2 | 3574 | ||
| AK313833.1 | - | 6 | 864 | |||
| AL110102.1 | - | 5 | 570 | |||
| AV711990.1 | - | 5 | 498 | |||
| BC004892.2 | - | 6 | 833 | 3505241 | ||
| BT009790.1 | - | 6 | 772 | |||
| X78669.1 | - | 6 | 838 | |||