Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zyg11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033634.3 | NP_001028806.2 | 3 | 535 | ||
GenBank mRNA | AK145130.1 | - | 3 | 508 | ||
AK148335.1 | - | 3 | 534 | |||
AK162646.1 | - | 3 | 498 | |||
AK166031.1 | - | 3 | 515 | |||
AK166174.1 | - | 3 | 515 | |||
AK166278.1 | - | 3 | 525 | |||
BC117977.2 | - | 3 | 499 | 40099428 | ||
BC117978.2 | - | 3 | 499 | 40099429 | ||
BC139383.1 | - | 3 | 502 | 8862405 | ||
BC139384.1 | - | 3 | 502 | 8862406 |