Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS3 SIRNA Assays ›
          Gene Symbol:
          RPS3
          Gene Name:
          ribosomal protein S3
          Gene Aliases:
          S3
          Chromosome Location:
          Chr.11: 75399491 - 75422302 on Build GRCh38
          UniGene ID:
          Hs.546286
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s12257

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPS3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005.4 NP_000996.2 2 118
          NM_001256802.1 NP_001243731.1 2 118
          NM_001260506.1 NP_001247435.1 2 118
          GenBank mRNA AB062288.1 - 2 54
          AK129794.1 - 2 90
          AK130960.1 - 2 91
          AK223048.1 - 2 67
          AK313051.1 - 2 87
          BC003137.1 - 2 67 2906158
          BC003577.1 - 2 59
          BC013196.1 - 2 59
          BC013231.1 - 2 79
          BC029981.1 - 1 12
          BC034149.1 - 2 91 4665438
          BC071917.1 - 2 70 6297012
          BC100284.1 - 2 79 6160514
          BM831460.1 - 2 118
          BM928330.1 - 2 100
          BQ645330.1 - 78
          S42658.1 - 2 69
          U14990.1 - 2 61
          U14991.1 - 2 61
          U14992.1 - 2 61
          X55715.1 - 2 83

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001009239, X51536, BC088450.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          DNA repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          apoptotic process
          cellular response to DNA damage stimulus
          response to oxidative stress
          chromosome segregation
          mitotic nuclear division
          positive regulation of gene expression
          negative regulation of translation
          viral transcription
          positive regulation of microtubule polymerization
          negative regulation of protein ubiquitination
          positive regulation of endodeoxyribonuclease activity
          DNA damage response, detection of DNA damage
          regulation of apoptotic process
          positive regulation of JUN kinase activity
          negative regulation of DNA repair
          positive regulation of DNA repair
          spindle assembly
          cell division
          response to TNF agonist
          cellular response to hydrogen peroxide
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          positive regulation of DNA N-glycosylase activity
          positive regulation of base-excision repair
          positive regulation of apoptotic signaling pathway
          positive regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
          DNA binding
          damaged DNA binding
          RNA binding
          mRNA binding
          structural constituent of ribosome
          DNA-(apurinic or apyrimidinic site) lyase activity
          endodeoxyribonuclease activity
          protein binding
          microtubule binding
          transcription factor binding
          tubulin binding
          DNA N-glycosylase activity
          enzyme binding
          kinase binding
          protein kinase binding
          Hsp70 protein binding
          SUMO binding
          oxidized purine DNA binding
          oxidized pyrimidine DNA binding
          ubiquitin-like protein conjugating enzyme binding
          poly(A) RNA binding
          protein kinase A binding
          iron-sulfur cluster binding
          Hsp90 protein binding
          small ribosomal subunit rRNA binding
          supercoiled DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0