Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gys2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145572.2 | NP_663547.2 | 4, 5 | 993 | ||
GenBank mRNA | AK138992.1 | - | 4, 5 | 701 | ||
AK149391.1 | - | 4, 5 | 717 | |||
BC158081.1 | - | 4, 5 | 854 | 9007373 |