Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SCNN1D SIRNA Assays ›
Gene Symbol:
SCNN1D
Gene Name:
sodium channel epithelial 1 delta subunit
Gene Aliases:
ENaCd, ENaCdelta, SCNED, dNaCh
Chromosome Location:
Chr.1: 1280415 - 1292029 on Build GRCh38
UniGene ID:
Hs.512681
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12549
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SCNN1D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130413.3
NP_001123885.2
17
2228
NR_037668.2
16
2134
XM_011541899.2
XP_011540201.1
17
2898
XM_011541901.2
XP_011540203.1
18
2420
XM_011541902.2
XP_011540204.1
18
2396
XM_011541905.2
XP_011540207.1
17
2324
XM_011541906.2
XP_011540208.1
14
2658
XM_011541908.2
XP_011540210.1
17
2595
XM_011541920.2
XP_011540222.1
16
2418
XM_011541925.2
XP_011540227.1
15
1977
XM_011541929.2
XP_011540231.1
13
2786
XM_011541932.2
XP_011540234.1
14
1883
XM_011541933.2
XP_011540235.1
14
1849
XM_017002037.1
XP_016857526.1
16
2895
XM_017002038.1
XP_016857527.1
15
2748
XM_017002039.1
XP_016857528.1
17
2273
XM_017002040.1
XP_016857529.1
17
2135
XM_017002041.1
XP_016857530.1
18
2123
XM_017002042.1
XP_016857531.1
17
2114
XM_017002043.1
XP_016857532.1
17
2090
XM_017002044.1
XP_016857533.1
16
2039
GenBank mRNA
AK093239.1
-
14
1858
AK093372.1
-
11
983
AK127357.1
-
14
2654
AK131558.1
-
18
2554
AK315288.1
-
13
1705
BC036752.1
-
13
1677
BC040882.1
-
16
2098
BC125074.1
-
13
1627
40117641
BC125075.1
-
12
1729
40117643
DQ898175.1
-
13
1602
DQ898176.1
-
14
1708
EU489064.1
-
12
1510
U38254.1
-
12
1510
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
ion transmembrane transport
sodium ion transmembrane transport
response to stimulus
sensory perception of taste
protein binding
ligand-gated sodium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline