Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA8 SIRNA Assays ›
          Gene Symbol:
          Hspa8
          Gene Name:
          heat shock protein 8
          Gene Aliases:
          2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
          Chromosome Location:
          Chr.9: 40801273 - 40805199 on Build GRCm38
          UniGene ID:
          Mm.290774
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s125649

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hspa8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031165.4 NP_112442.2 3 466
          GenBank mRNA AK004608.1 - 2 755
          AK035286.1 - 3 464
          AK075935.1 - 3 463
          AK081701.1 - 3 464
          AK098074.1 - 3 463
          AK145416.1 - 3 386
          AK145579.1 - 3 460
          AK146547.1 - 3 467
          AK146708.1 - 3 461
          AK146985.1 - 3 464
          AK147864.1 - 3 464
          AK149954.1 - 3 461
          AK150474.1 - 3 458
          AK150498.1 - 3 458
          AK150701.1 - 3 468
          AK150958.1 - 3 461
          AK151041.1 - 3 464
          AK151065.1 - 3 464
          AK151127.1 - 3 467
          AK151287.1 - 3 464
          AK151435.1 - 3 464
          AK151516.1 - 3 470
          AK151537.1 - 3 461
          AK151741.1 - 3 464
          AK151775.1 - 3 466
          AK151808.1 - 3 467
          AK151865.1 - 3 464
          AK151892.1 - 3 466
          AK151948.1 - 3 467
          AK151997.1 - 3 464
          AK152315.1 - 3 467
          AK152598.1 - 3 464
          AK152697.1 - 3 464
          AK152703.1 - 3 464
          AK152768.1 - 3 467
          AK152803.1 - 3 467
          AK153032.1 - 3 466
          AK153834.1 - 3 464
          AK157831.1 - 3 464
          AK159479.1 - 3 464
          AK162775.1 - 3 464
          AK163987.1 - 3 464
          AK164000.1 - 3 464
          AK166545.1 - 3 466
          AK166643.1 - 3 478
          AK166721.1 - 3 461
          AK166767.1 - 3 464
          AK166776.1 - 3 467
          AK166808.1 - 3 464
          AK166830.1 - 3 462
          AK166846.1 - 3 464
          AK166861.1 - 3 467
          AK166873.1 - 3 464
          AK166908.1 - 3 467
          AK166910.1 - 3 464
          AK166913.1 - 3 464
          AK166933.1 - 3 464
          AK167043.1 - 3 466
          AK167121.1 - 3 467
          AK167122.1 - 3 464
          AK167134.1 - 3 462
          AK167163.1 - 3 466
          AK167218.1 - 3 464
          AK167229.1 - 3 465
          AK167254.1 - 3 465
          AK167414.1 - 3 464
          AK167845.1 - 3 467
          AK167910.1 - 3 467
          AK168435.1 - 3 467
          AK168492.1 - 3 466
          AK168519.1 - 3 464
          AK168542.1 - 3 466
          AK168577.1 - 3 464
          AK168625.1 - 3 464
          AK168654.1 - 3 464
          AK168711.1 - 3 464
          AK168750.1 - 3 467
          AK168776.1 - 3 467
          AK168887.1 - 3 466
          AK168934.1 - 3 467
          AK169093.1 - 3 464
          AK169179.1 - 3 464
          AK169236.1 - 3 464
          AK169293.1 - 3 478
          AK169329.1 - 3 467
          AK169594.1 - 3 463
          AK171434.1 - 3 465
          AK185576.1 - 2 225
          AK188190.1 - 1 172
          AK188441.1 - 1 172
          AK190603.1 - 1 172
          AK204952.1 - 1 172
          AK206788.1 - 1 172
          AK212540.1 - 1 172
          AK218563.1 - 1 172
          BC006722.1 - 3 488 3711399
          BC066191.1 - 3 420 30467052
          BC085486.1 - 3 452 30540914
          BC089322.1 - 3 445 30538640
          BC089457.1 - 3 423 30628505
          BC094900.1 - 3 437 30063483
          BC106169.1 - 1 144
          BC106193.1 - 3 448 6329283
          M19141.1 - 3 450
          U27129.1 - 3 415

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_153201, NM_006597, AF352832, AK222628, AK096100, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          protein folding
          RNA splicing
          protein refolding
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone mediated protein folding requiring cofactor
          regulation of cell cycle
          chaperone-mediated protein folding
          clathrin coat disassembly
          negative regulation of fibril organization
          nucleotide binding
          G-protein coupled receptor binding
          receptor binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ATPase activity, coupled
          ADP binding
          poly(A) RNA binding
          unfolded protein binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0