Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSPA8 SIRNA Assays ›
Gene Symbol:
Hspa8
Gene Name:
heat shock protein 8
Gene Aliases:
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location:
Chr.9: 40801273 - 40805199 on Build GRCm38
UniGene ID:
Mm.290774
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s125651
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hspa8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031165.4
NP_112442.2
5
779
GenBank mRNA
AK004608.1
-
4
1068
AK035286.1
-
5
777
AK075935.1
-
5
776
AK081701.1
-
5
778
AK098074.1
-
5
776
AK145416.1
-
5
699
AK145579.1
-
5
773
AK146547.1
-
5
780
AK146708.1
-
5
774
AK146985.1
-
5
777
AK147864.1
-
5
777
AK149954.1
-
5
774
AK150474.1
-
5
771
AK150498.1
-
5
771
AK150701.1
-
5
781
AK150958.1
-
5
774
AK151041.1
-
5
777
AK151065.1
-
5
777
AK151127.1
-
5
780
AK151287.1
-
5
777
AK151435.1
-
5
777
AK151516.1
-
5
783
AK151537.1
-
5
774
AK151741.1
-
5
777
AK151775.1
-
5
779
AK151808.1
-
5
780
AK151865.1
-
5
777
AK151892.1
-
5
779
AK151948.1
-
5
780
AK151997.1
-
5
777
AK152315.1
-
5
780
AK152598.1
-
5
777
AK152697.1
-
5
777
AK152703.1
-
5
777
AK152768.1
-
5
780
AK152803.1
-
5
780
AK153032.1
-
5
779
AK153834.1
-
5
777
AK157831.1
-
5
777
AK159479.1
-
5
777
AK162775.1
-
5
777
AK163987.1
-
5
777
AK164000.1
-
5
777
AK166545.1
-
5
779
AK166643.1
-
5
791
AK166721.1
-
5
774
AK166767.1
-
5
777
AK166776.1
-
5
780
AK166808.1
-
5
777
AK166830.1
-
5
775
AK166846.1
-
5
777
AK166861.1
-
5
780
AK166873.1
-
5
777
AK166908.1
-
5
780
AK166910.1
-
5
777
AK166913.1
-
5
777
AK166933.1
-
5
777
AK167043.1
-
5
779
AK167121.1
-
5
780
AK167122.1
-
5
777
AK167134.1
-
5
775
AK167163.1
-
5
779
AK167218.1
-
5
777
AK167229.1
-
5
778
AK167254.1
-
5
778
AK167414.1
-
5
777
AK167845.1
-
5
780
AK167910.1
-
5
780
AK168435.1
-
5
780
AK168492.1
-
5
779
AK168511.1
-
3
315
AK168519.1
-
5
777
AK168542.1
-
5
779
AK168577.1
-
5
777
AK168625.1
-
5
777
AK168654.1
-
5
777
AK168711.1
-
5
777
AK168750.1
-
5
780
AK168776.1
-
5
780
AK168887.1
-
5
779
AK168934.1
-
5
780
AK169093.1
-
5
777
AK169179.1
-
5
777
AK169236.1
-
5
777
AK169293.1
-
5
791
AK169329.1
-
5
780
AK169594.1
-
5
776
AK171434.1
-
5
778
AK178143.1
-
2
258
AK184919.1
-
2
258
AK190509.1
-
2
258
AK192182.1
-
2
259
AK192272.1
-
2
258
AK193850.1
-
2
258
AK195240.1
-
2
259
AK198438.1
-
2
258
AK198532.1
-
3
319
AK199142.1
-
2
258
AK199143.1
-
2
259
AK199178.1
-
2
258
AK199410.1
-
2
291
AK199504.1
-
2
259
AK200099.1
-
2
259
AK201767.1
-
2
258
AK202859.1
-
2
258
AK203911.1
-
2
259
AK204890.1
-
2
258
AK205054.1
-
2
259
AK205187.1
-
2
258
AK206134.1
-
2
259
AK206953.1
-
2
258
AK209289.1
-
2
258
AK210012.1
-
3
324
AK211519.1
-
2
258
AK212081.1
-
3
322
AK212914.1
-
2
278
AK214003.1
-
3
305
AK216159.1
-
2
258
AK216732.1
-
2
258
AK216875.1
-
2
259
BC006722.1
-
5
801
3711399
BC066191.1
-
5
733
30467052
BC085486.1
-
5
765
30540914
BC089322.1
-
5
758
30538640
BC089457.1
-
5
736
30628505
BC094900.1
-
5
693
30063483
BC106169.1
-
3
457
BC106193.1
-
5
761
6329283
U27129.1
-
5
728
X05837.1
-
1
131
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_153201, NM_006597, AF352832, AK222628, AK096100, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
protein refolding
negative regulation of transcription, DNA-templated
ATP metabolic process
positive regulation of mRNA splicing, via spliceosome
chaperone mediated protein folding requiring cofactor
regulation of cell cycle
chaperone-mediated protein folding
clathrin coat disassembly
negative regulation of fibril organization
nucleotide binding
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
MHC class II protein complex binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
ADP binding
poly(A) RNA binding
unfolded protein binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0