Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP3K5 SIRNA Assays ›
Gene Symbol:
Map3k5
Gene Name:
mitogen-activated protein kinase kinase kinase 5
Gene Aliases:
7420452D20Rik, ASK, ASK1, MAPKKK5, Mekk5
Chromosome Location:
Chr.10: 19934513 - 20142750 on Build GRCm38
UniGene ID:
Mm.6595
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s126922
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Map3k5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008580.4
NP_032606.4
30
4605
GenBank mRNA
AK033726.1
-
6
1037
AK078668.1
-
16
2401
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008758622.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
activation of MAPK activity
positive regulation of protein phosphorylation
immune system process
protein phosphorylation
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
JNK cascade
intrinsic apoptotic signaling pathway in response to oxidative stress
regulation of cell death
phosphorylation
response to endoplasmic reticulum stress
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of JUN kinase activity
innate immune response
positive regulation of JNK cascade
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to hydrogen peroxide
programmed necrotic cell death
positive regulation of neuron death
cellular response to reactive nitrogen species
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
cysteine-type endopeptidase activator activity involved in apoptotic process
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
protein phosphatase binding
protein homodimerization activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-gh6bl:80/100.66.76.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0