Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078166.1 | NP_001071634.1 | 3 | 742 | ||
NM_006924.4 | NP_008855.1 | 3 | 742 | |||
NR_034041.1 | 3 | 742 | ||||
GenBank mRNA | AB062124.1 | - | 3 | 623 | ||
AB209558.1 | - | 3 | 629 | |||
AK225711.1 | - | 3 | 620 | |||
AK289933.1 | - | 3 | 651 | |||
AK298527.1 | - | 2 | 324 | |||
AK312781.1 | - | 3 | 639 | |||
BC010264.1 | - | 3 | 568 | 2900101 | ||
BC033785.1 | - | 3 | 628 | 5166554 | ||
M69040.1 | - | 3 | 657 | |||
M72709.1 | - | 3 | 629 |