Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGT SIRNA Assays ›
          Gene Symbol:
          Agt
          Gene Name:
          angiotensinogen (serpin peptidase inhibitor, clade A, member 8)
          Gene Aliases:
          ANRT, Ang, AngII, PAT
          Chromosome Location:
          Chr.19: 57321594 - 57333462 on Build Rnor_6.0
          UniGene ID:
          Rn.6319
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s127415

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Agt
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_134432.2 NP_602308.1 2 604
          XM_008772597.1 XP_008770819.1 3 804
          GenBank mRNA BC078741.1 - 2 604 7128542
          BC087679.1 - 1 440 7318795

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_007428, BC028877, BC054387, AK139179, AK005354, and others.

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle rupture
          blood vessel development
          branching involved in ureteric bud morphogenesis
          kidney development
          regulation of systemic arterial blood pressure by circulatory renin-angiotensin
          angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure
          renal response to blood flow involved in circulatory renin-angiotensin regulation of systemic arterial blood pressure
          renin-angiotensin regulation of aldosterone production
          regulation of renal output by angiotensin
          brain renin-angiotensin system
          renal system process
          angiotensin-mediated drinking behavior
          positive regulation of extracellular matrix constituent secretion
          cellular sodium ion homeostasis
          cell-matrix adhesion
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          activation of phospholipase C activity
          activation of NF-kappaB-inducing kinase activity
          female pregnancy
          aging
          excretion
          establishment of blood-nerve barrier
          regulation of blood pressure
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to cold
          response to salt stress
          regulation of gene expression
          positive regulation of activation of JAK2 kinase activity
          positive regulation of endothelial cell migration
          positive regulation of cardiac muscle hypertrophy
          positive regulation of gene expression
          positive regulation of cholesterol esterification
          negative regulation of endopeptidase activity
          regulation of norepinephrine secretion
          positive regulation of phosphatidylinositol 3-kinase signaling
          artery smooth muscle contraction
          response to muscle activity involved in regulation of muscle adaptation
          negative regulation of angiogenesis
          extracellular matrix organization
          negative regulation of cell growth
          peristalsis
          positive regulation of cellular protein metabolic process
          positive regulation of superoxide anion generation
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of tissue remodeling
          catenin import into nucleus
          regulation of renal sodium excretion
          positive regulation of renal sodium excretion
          positive regulation of multicellular organism growth
          vasoconstriction
          vasodilation
          hormone metabolic process
          drinking behavior
          regulation of apoptotic process
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of fatty acid biosynthetic process
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of blood pressure
          positive regulation of transcription, DNA-templated
          positive regulation of vasodilation
          positive regulation of organ growth
          astrocyte activation
          fibroblast proliferation
          positive regulation of fibroblast proliferation
          regulation of long-term neuronal synaptic plasticity
          smooth muscle cell proliferation
          cytokine secretion
          regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          smooth muscle cell differentiation
          negative regulation of neurotrophin TRK receptor signaling pathway
          stress-activated MAPK cascade
          regulation of calcium ion transport
          regulation of transmission of nerve impulse
          cell growth involved in cardiac muscle cell development
          positive regulation of protein tyrosine kinase activity
          ERK1 and ERK2 cascade
          uterine smooth muscle contraction
          cellular response to mechanical stimulus
          positive regulation of branching involved in ureteric bud morphogenesis
          positive regulation of protein kinase C activity
          regulation of extracellular matrix assembly
          positive regulation of membrane hyperpolarization
          positive regulation of gap junction assembly
          regulation of cardiac conduction
          positive regulation of extrinsic apoptotic signaling pathway
          serine-type endopeptidase inhibitor activity
          hormone activity
          sodium channel regulator activity
          angiotensin receptor binding
          type 1 angiotensin receptor binding
          type 2 angiotensin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0