Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRL3D4 SIRNA Assays ›
          Gene Symbol:
          Prl3d4 LOC100911206 LOC103689989 LOC103690150 Prl3d1 Prl3d2
          Gene Name:
          prolactin family 3, subfamily d, member 4
          prolactin-3D1-like
          prolactin-3D4
          prolactin-3D1
          Prolactin family 3, subfamily d, member 1
          Prolactin family 3, subfamily d, member 2
          Gene Aliases:
          Csh1l1, Csh1v, Pl-Iv
          -
          -
          -
          Csh1, Pl-I, Pl-Im, Pl1, Rhco1, rPl-1
          Pl-1
          Chromosome Location:
          Chr.17: 39271883 - 39278716 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 81664409 - 81673264 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 42920887 - 42927745 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 39721993 - 39728391 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 39777255 - 39782877 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 42849469 - 42855494 on Build Rnor_6.0
          UniGene ID:
          Rn.1259
          -
          -
          Rn.112569
          Rn.112569
          Rn.198338
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s127618

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Prl3d4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033233.1 NP_150236.1 5 638
          XM_008771595.1 XP_008769817.1 5 620
          GenBank mRNA BC097302.1 - 5 664 7444366
          U32679.1 - 5 638
          Gene Symbol: LOC100911206
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006257188.2 XP_006257250.2 3 401
          Gene Symbol: LOC103689989
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771673.1 XP_008769895.1 620
          XM_008771674.1 XP_008769896.1 658
          Gene Symbol: LOC103690150
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771672.1 XP_008769894.1 658
          GenBank mRNA FQ228618.1 - 681
          Gene Symbol: Prl3d1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006253914.2 5 681
          GenBank mRNA FQ220053.1 - 685
          FQ220072.1 - 681
          FQ220274.1 - 683
          FQ220568.1 - 683
          FQ220587.1 - 681
          FQ220864.1 - 407
          FQ221231.1 - 683
          FQ222593.1 - 681
          FQ228787.1 - 681
          FQ229328.1 - 681
          FQ229537.1 - 680
          FQ229656.1 - 675
          FQ230119.1 - 681
          M55269.1 - 674
          Gene Symbol: Prl3d2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771671.1 XP_008769893.1 5 684
          GenBank mRNA BC161851.1 - 5 658 9088452
          D21103.1 - 5 620
          DQ329282.1 - 5 677
          DQ329283.1 - 5 677
          FQ145043.1 - 5 683
          FQ221365.1 - 5 681
          FQ223255.1 - 5 684
          FQ223518.1 - 5 681
          FQ229928.1 - 5 683
          FQ230035.1 - 5 681

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          activation of Janus kinase activity
          prolactin receptor binding
          hormone activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lfs6t:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0