Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITSN1 SIRNA Assays ›
Gene Symbol:
ITSN1
Gene Name:
intersectin 1
Gene Aliases:
ITSN, SH3D1A, SH3P17
Chromosome Location:
Chr.21: 33642400 - 33899861 on Build GRCh38
UniGene ID:
Hs.160324
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12794
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ITSN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001132.1
NP_001001132.1
9
1010
NM_003024.2
NP_003015.2
9
1010
XM_005261026.2
XP_005261083.1
9
1088
XM_005261027.1
XP_005261084.1
9
1088
XM_005261031.1
XP_005261088.1
9
1088
XM_011529691.1
XP_011527993.1
9
977
XM_011529692.1
XP_011527994.1
9
977
XM_011529693.2
XP_011527995.1
9
1088
XM_017028428.1
XP_016883917.1
10
1008
XM_017028429.1
XP_016883918.1
9
977
XM_017028430.1
XP_016883919.1
9
977
XM_017028431.1
XP_016883920.1
9
1088
XM_017028432.1
XP_016883921.1
9
1088
XM_017028433.1
XP_016883922.1
9
1088
XM_017028434.1
XP_016883923.1
9
1088
XM_017028435.1
XP_016883924.1
9
977
XM_017028436.1
XP_016883925.1
9
977
XM_017028437.1
XP_016883926.1
5
544
XM_017028438.1
XP_016883927.1
9
977
XM_017028439.1
XP_016883928.1
9
1088
XM_017028440.1
XP_016883929.1
9
977
XM_017028441.1
XP_016883930.1
9
1088
GenBank mRNA
AF064243.1
-
9
848
AF064244.1
-
9
848
AF114487.1
-
9
1010
AF114488.1
-
9
1010
AK027846.1
-
9
977
AK074554.1
-
9
946
AK075290.1
-
9
1005
AK300274.1
-
9
985
BC013578.1
-
9
873
BC020269.1
-
9
923
BC039036.1
-
9
1005
BC058925.1
-
9
1037
BC073744.1
-
9
952
BC116185.1
-
9
821
40073780
BC116186.2
-
9
821
40073781
BC117560.1
-
9
990
7469144
BX538175.1
-
4
309
DQ340865.1
-
8
738
DQ386455.1
-
8
761
DQ679754.1
-
9
827
DQ679755.1
-
9
827
DQ679756.1
-
10
716
DQ679757.1
-
9
827
EU107386.1
-
9
716
EU117382.1
-
8
743
EU120733.1
-
8
743
EU120734.1
-
8
743
EU120735.1
-
9
716
EU120736.1
-
9
716
EU140799.1
-
9
827
EU140800.1
-
8
632
EU152331.1
-
8
743
EU191901.1
-
8
666
GU939616.1
-
4
432
GU939617.1
-
4
432
GU939621.1
-
4
734
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
regulation of Rho protein signal transduction
positive regulation of phosphorylation
positive regulation of apoptotic process
negative regulation of neuron apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
synaptic vesicle endocytosis
regulation of small GTPase mediated signal transduction
positive regulation of protein kinase B signaling
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
calcium ion binding
protein binding
kinase activator activity
protein complex scaffold
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0