Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INS2 SIRNA Assays ›
          Gene Symbol:
          Ins2 Ins1
          Gene Name:
          insulin 2
          insulin 1
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.1: 215856967 - 215858034 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 272799784 - 272800351 on Build Rnor_6.0
          UniGene ID:
          Rn.989
          Rn.962
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s127966

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ins2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019130.2 NP_062003.1 2 133
          GenBank mRNA BP472188.1 - 2 108
          BP485158.1 - 2 132
          Gene Symbol: Ins1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019129.3 NP_062002.1 2 133
          GenBank mRNA BP465191.1 - 1 85
          BP502578.1 - 2 153
          CB702626.1 - 2 120

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_008386, AK148541, BC145868, AK007345, AK007482, and others.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          glucose metabolic process
          ER overload response
          myoblast fusion
          biological_process
          insulin receptor signaling pathway
          lipid catabolic process
          regulation of protein binding
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of peptidyl-tyrosine phosphorylation
          canonical Wnt signaling pathway
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          MAPK cascade
          negative regulation of acute inflammatory response
          regulation of cellular amino acid metabolic process
          acute-phase response
          G-protein coupled receptor signaling pathway
          positive regulation of cell proliferation
          response to organic substance
          positive regulation of phosphatidylinositol 3-kinase signaling
          glucose transport
          regulation of transmembrane transporter activity
          activation of protein kinase B activity
          positive regulation of cellular protein metabolic process
          negative regulation of protein oligomerization
          regulation of protein localization
          negative regulation of NAD(P)H oxidase activity
          response to cytokine
          wound healing
          negative regulation of protein catabolic process
          glucose homeostasis
          positive regulation of MAPK cascade
          response to peptide hormone
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          negative regulation of glycogen catabolic process
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          negative regulation of proteolysis
          negative regulation of fatty acid metabolic process
          positive regulation of glucose import
          positive regulation of insulin receptor signaling pathway
          alpha-beta T cell activation
          regulation of protein secretion
          negative regulation of protein secretion
          positive regulation of cytokine secretion
          negative regulation of lipid catabolic process
          positive regulation of NF-kappaB transcription factor activity
          response to cAMP
          positive regulation of protein kinase B signaling
          fatty acid homeostasis
          negative regulation of respiratory burst involved in inflammatory response
          positive regulation of respiratory burst
          positive regulation of protein localization to nucleus
          negative regulation of feeding behavior
          protease binding
          hormone activity
          protein complex binding
          insulin receptor binding
          insulin-like growth factor receptor binding
          identical protein binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qq54g:80/100.66.72.153:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0