Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017029.2 | NP_058725.1 | 3 | 2457 | ||
XM_008770736.1 | XP_008768958.1 | 4 | 2794 | |||
GenBank mRNA | M18628.1 | - | 3 | 2432 | ||
Z12152.1 | - | 3 | 2460 |