Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDGFRB SIRNA Assays ›
          Gene Symbol:
          Pdgfrb
          Gene Name:
          platelet derived growth factor receptor, beta polypeptide
          Gene Aliases:
          PDGFR-1
          Chromosome Location:
          Chr.18: 56364587 - 56406381 on Build Rnor_6.0
          UniGene ID:
          Rn.98311
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s128177

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pdgfrb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031525.1 NP_113713.1 21 3258
          XM_006254789.2 XP_006254851.1 21 3348
          XM_008772084.1 XP_008770306.1 20 2899
          GenBank mRNA AF359356.1 - 11 1311
          AY090783.1 - 21 3258
          DQ979389.1 - 15 1997

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_008809, NM_001146268, DQ979390, AK158145, AK134785, and others.

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          kidney development
          tissue homeostasis
          glycosaminoglycan biosynthetic process
          nitrogen compound metabolic process
          signal transduction
          positive regulation of cell proliferation
          response to toxic substance
          positive regulation of phospholipase C activity
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to organic cyclic compound
          positive regulation of smooth muscle cell migration
          phosphorylation
          cell migration
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          adrenal gland development
          positive regulation of cell migration
          response to estradiol
          positive regulation of phosphoprotein phosphatase activity
          response to retinoic acid
          regulation of actin cytoskeleton organization
          positive regulation of collagen biosynthetic process
          response to lipid
          response to fluid shear stress
          cell migration involved in vasculogenesis
          intracellular signal transduction
          metanephric mesenchymal cell migration
          platelet-derived growth factor receptor-beta signaling pathway
          positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
          aorta morphogenesis
          cellular response to platelet-derived growth factor stimulus
          positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
          wound healing
          response to hydrogen peroxide
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          response to estrogen
          positive regulation of mitotic nuclear division
          phosphatidylinositol metabolic process
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of smooth muscle cell proliferation
          skeletal system morphogenesis
          smooth muscle tissue development
          inner ear development
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of chemotaxis
          cardiac myofibril assembly
          response to hyperoxia
          cell chemotaxis
          cell migration involved in coronary angiogenesis
          retina vasculature development in camera-type eye
          positive regulation of ERK1 and ERK2 cascade
          smooth muscle cell chemotaxis
          metanephric mesenchyme development
          metanephric glomerular mesangial cell proliferation involved in metanephros development
          metanephric glomerulus morphogenesis
          metanephric glomerular capillary formation
          metanephric comma-shaped body morphogenesis
          metanephric S-shaped body morphogenesis
          positive regulation of calcium ion import
          positive regulation of reactive oxygen species metabolic process
          positive regulation of DNA biosynthetic process
          protein kinase activity
          protein tyrosine kinase activity
          signal transducer activity
          platelet-derived growth factor-activated receptor activity
          platelet-derived growth factor beta-receptor activity
          receptor binding
          platelet-derived growth factor receptor binding
          protein binding
          ATP binding
          kinase activity
          protein kinase binding
          vascular endothelial growth factor binding
          phosphatidylinositol 3-kinase binding
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline