Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SERPINA1 SIRNA Assays ›
Gene Symbol:
Serpina1
Gene Name:
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
Gene Aliases:
AAT, Pi, Spi1
Chromosome Location:
Chr.6: 127610241 - 127632265 on Build Rnor_6.0
UniGene ID:
Rn.1419
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128203
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Serpina1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022519.2
NP_071964.2
3
717
XM_006240456.1
XP_006240518.1
2
574
GenBank mRNA
BC078824.1
-
3
717
7131535
D00675.1
-
2
520
FQ209494.1
-
1
31
FQ209517.1
-
1
31
FQ209686.1
-
1
31
FQ209716.1
-
1
31
FQ209779.1
-
1
31
FQ209814.1
-
1
31
FQ209897.1
-
1
31
FQ209914.1
-
1
31
FQ210169.1
-
1
31
FQ210655.1
-
2
510
FQ210685.1
-
1
31
FQ210775.1
-
1
31
FQ210989.1
-
1
31
FQ210996.1
-
1
31
FQ211012.1
-
1
31
FQ211042.1
-
1
31
FQ211203.1
-
1
31
FQ211353.1
-
1
31
FQ218226.1
-
1
31
FQ218241.1
-
1
31
FQ218252.1
-
1
31
FQ218261.1
-
1
31
FQ218411.1
-
2
649
FQ218460.1
-
1
31
FQ218555.1
-
1
31
FQ218567.1
-
1
31
FQ218586.1
-
1
31
FQ218654.1
-
1
31
FQ218694.1
-
1
31
FQ218710.1
-
1
31
FQ218855.1
-
1
31
FQ218857.1
-
1
31
FQ218947.1
-
1
31
FQ218955.1
-
1
31
FQ218993.1
-
1
31
FQ219224.1
-
1
31
FQ219252.1
-
1
31
FQ219268.1
-
1
31
FQ219276.1
-
1
31
FQ219303.1
-
1
31
FQ219441.1
-
1
31
FQ219458.1
-
1
31
FQ219621.1
-
1
31
FQ219652.1
-
1
31
FQ219732.1
-
1
31
FQ219741.1
-
2
511
FQ219765.1
-
1
31
M32247.1
-
1
457
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
acute-phase response
inflammatory response
response to inorganic substance
response to lead ion
response to organic cyclic compound
response to estradiol
response to lipopolysaccharide
response to methanol
response to triglyceride
response to cytokine
response to chromate
negative regulation of serine-type endopeptidase activity
glycoprotein binding
protease binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bfkrn:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0