Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SHMT2 SIRNA Assays ›
          Gene Symbol:
          SHMT2
          Gene Name:
          serine hydroxymethyltransferase 2
          Gene Aliases:
          GLYA, HEL-S-51e, SHMT
          Chromosome Location:
          Chr.12: 57229573 - 57234935 on Build GRCh38
          UniGene ID:
          Hs.741179
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s12825

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SHMT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166356.1 NP_001159828.1 8 1047
          NM_001166357.1 NP_001159829.1 8 1139
          NM_001166358.1 NP_001159830.1 8 1038
          NM_001166359.1 NP_001159831.1 8 931
          NM_005412.5 NP_005403.2 8 1077
          NR_029415.1 7 879
          NR_029416.1 7 958
          NR_029417.1 7 997
          NR_048562.1 7 1087
          XM_011538675.2 XP_011536977.1 8 1017
          XM_011538676.2 XP_011536978.1 8 1008
          XM_011538677.2 XP_011536979.1 7 918
          XM_011538678.2 XP_011536980.1 7 900
          GenBank mRNA AK055053.1 - 4 2063
          AK223555.1 - 8 939
          AK295941.1 - 7 933
          AK296183.1 - 8 891
          AK297173.1 - 7 839
          AK298231.1 - 7 732
          AK301339.1 - 8 804
          AK301443.1 - 7 901
          AK303827.1 - 7 753
          AK315916.1 - 8 1131
          BC011911.2 - 8 881 4139001
          BC013677.2 - 8 941 3863368
          BC025355.1 - 5 464
          BC032584.1 - 8 920 5499481
          BC044211.1 - 8 917 5521082
          BC091501.1 - 7 801
          BT006866.1 - 8 871
          BX647711.1 - 8 998
          BX648847.1 - 6 479
          EU668344.1 - 8 881
          KU178333.1 - 8 871
          KU178334.1 - 7 507
          L11932.1 - 7 782

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_028230, NM_001252316, BC004825, AK012355, AK051207, and others.

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycine metabolic process
          L-serine metabolic process
          L-serine biosynthetic process
          one-carbon metabolic process
          positive regulation of cell proliferation
          glycine biosynthetic process from serine
          response to type I interferon
          tetrahydrofolate interconversion
          tetrahydrofolate metabolic process
          folic acid metabolic process
          protein tetramerization
          protein homotetramerization
          protein K63-linked deubiquitination
          chromatin binding
          glycine hydroxymethyltransferase activity
          protein binding
          L-allo-threonine aldolase activity
          amino acid binding
          pyridoxal phosphate binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0