Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC299282 SIRNA Assays ›
          Gene Symbol:
          LOC299282 LOC100909605 RGD1565462 Serpina3c
          Gene Name:
          Serine protease inhibitor
          serine protease inhibitor A3F-like
          similar to BC049975 protein
          serine (or cysteine) proteinase inhibitor, clade A, member 3C
          Gene Aliases:
          Ab1-021, CPi-23, SPI1, Serpina3l, Spin1, Spin2a
          -
          -
          CPi-21, GHR-P63, Kbp, Klkbp, SPI-2, SPI-2.3, SPI2, Serpina3k, Spin2, Spin2b
          Chromosome Location:
          Chr.6: 128046780 - 128062241 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 129052483 - 129058971 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 127927716 - 127934204 on Build Rnor_6.0
          Chromosome Location:
          Chr.6: 127996162 - 128003435 on Build Rnor_6.0
          UniGene ID:
          Rn.97597
          -
          -
          Rn.91257
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s128438

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC299282
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC088096.1 - 3 787 7372979
          D00752.1 - 3 770
          FQ209483.1 - 3 783
          FQ210703.1 - 3 786
          FQ218472.1 - 3 788
          M15917.1 - 2 487
          X13149.1 - 2 84
          X16357.1 - 3 783
          Gene Symbol: LOC100909605
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006240519.2 XP_006240581.2 761
          Gene Symbol: RGD1565462
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008764903.1 XP_008763125.1 3 761
          Gene Symbol: Serpina3c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012657.2 NP_036789.2 3 778
          XM_006240458.2 XP_006240520.1 3 821
          GenBank mRNA BC062236.1 - 3 782 6888893
          CO560755.1 - 2 273
          D00751.1 - 3 731
          FQ209380.1 - 3 779
          FQ209577.1 - 3 780
          FQ210286.1 - 3 783
          FQ218085.1 - 3 780
          FQ218647.1 - 2 455
          FQ219296.1 - 3 783
          FQ219312.1 - 3 780
          FQ219600.1 - 3 779
          M14320.1 - 2 276
          M15916.1 - 3 743
          X05348.1 - 3 765
          X16358.1 - 3 779

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001033335, NM_009253, NM_011458, NM_001168294, NM_001168295, and others.

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          response to cytokine
          response to peptide hormone
          negative regulation of endothelial cell proliferation
          negative regulation of angiogenesis
          positive regulation of apoptotic process
          serine-type endopeptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hdb68:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0