Search
Search

ST3GAL2| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006927.3 | NP_008858.1 | 4 | 1741 | ||
| GenBank mRNA | AK127322.1 | - | 3 | 2706 | ||
| AK313020.1 | - | 3 | 872 | |||
| BC036777.1 | - | 3 | 640 | 5750330 | ||
| U63090.1 | - | 3 | 1278 | |||
| X96667.1 | - | 3 | 894 | |||