Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030989.3 | NP_112251.2 | 2 | 191 | ||
XM_006246594.2 | XP_006246656.1 | 3 | 620 | |||
XM_006246595.2 | XP_006246657.1 | 2 | 977 | |||
XM_008767773.1 | XP_008765995.1 | 2 | 198 | |||
GenBank mRNA | AF209191.1 | - | 1 | 83 | ||
AY009504.1 | - | 1 | 60 | |||
BC081788.1 | - | 2 | 120 | 7193583 | ||
BC098663.1 | - | 2 | 149 | 7384467 | ||
CV126174.1 | - | 2 | 191 | |||
FQ232659.1 | - | 2 | 175 | |||
U90328.1 | - | 1 | 60 | |||
X13058.1 | - | 1 | 83 |