Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030989.3 | NP_112251.2 | 6 | 720 | ||
XM_006246594.2 | XP_006246656.1 | 7 | 1149 | |||
XM_006246595.2 | XP_006246657.1 | 6 | 1506 | |||
XM_008767773.1 | XP_008765995.1 | 6 | 727 | |||
GenBank mRNA | AF209191.1 | - | 5 | 612 | ||
BC081788.1 | - | 6 | 649 | 7193583 | ||
BC098663.1 | - | 6 | 678 | 7384467 | ||
FQ232659.1 | - | 6 | 704 | |||
U90328.1 | - | 5 | 589 | |||
X13058.1 | - | 5 | 612 |