Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030989.3 | NP_112251.2 | 5 | 513 | ||
XM_006246594.2 | XP_006246656.1 | 6 | 942 | |||
XM_006246595.2 | XP_006246657.1 | 5 | 1299 | |||
XM_008767773.1 | XP_008765995.1 | 5 | 520 | |||
GenBank mRNA | AF209191.1 | - | 4 | 405 | ||
BC081788.1 | - | 5 | 442 | 7193583 | ||
BC098663.1 | - | 5 | 471 | 7384467 | ||
CV126174.1 | - | 5 | 513 | |||
FQ232659.1 | - | 5 | 497 | |||
U90328.1 | - | 4 | 382 | |||
X13058.1 | - | 4 | 405 |