Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Klrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012745.2 | NP_036877.1 | 7 | 860 | ||
XM_006237078.2 | XP_006237140.1 | 7 | 909 | |||
GenBank mRNA | AF009133.1 | - | 7 | 617 | ||
BC086393.1 | - | 7 | 860 | 7303354 |