Search
Search

Mfge8| Gene Symbol: Mfge8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040186.2 | NP_001035276.1 | 10 | 1442 | ||
| NM_012811.3 | NP_036943.1 | 9 | 1274 | |||
| GenBank mRNA | BC085754.1 | - | 9 | 1227 | 7193796 | |
| D84068.1 | - | 9 | 1234 | |||
| DQ455823.1 | - | 10 | 1395 | |||