Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CST3 SIRNA Assays ›
          Gene Symbol:
          Cst3
          Gene Name:
          cystatin C
          Gene Aliases:
          CYSC
          Chromosome Location:
          Chr.3: 143219671 - 143223544 on Build Rnor_6.0
          UniGene ID:
          Rn.106351
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s129432

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cst3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012837.1 NP_036969.1 2 351
          GenBank mRNA BC087591.1 - 2 291 7307694
          CB314369.1 - 2 123
          CB608544.1 - 2 334
          CB692474.1 - 2 361
          FQ211221.1 - 2 304
          FQ211526.1 - 2 304
          FQ211840.1 - 2 304
          FQ211941.1 - 2 304
          FQ212057.1 - 2 333
          FQ212112.1 - 2 304
          FQ212277.1 - 2 304
          FQ212367.1 - 2 304
          FQ212374.1 - 2 304
          FQ214337.1 - 2 304
          FQ218053.1 - 2 335
          FQ221232.1 - 2 304
          FQ221782.1 - 2 304
          FQ222268.1 - 2 304
          FQ222820.1 - 2 304
          FQ223224.1 - 2 304
          FQ224103.1 - 2 304
          FQ228332.1 - 2 301
          FQ228399.1 - 2 304
          FQ228646.1 - 2 304
          FQ234324.1 - 2 335
          X16957.1 - 2 245

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_009976, AF311741, AK166430, AK131728, AK153533, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          response to hypoxia
          cell activation
          apoptotic process
          defense response
          response to oxidative stress
          brain development
          salivary gland development
          embryo implantation
          positive regulation of cell proliferation
          male gonad development
          response to toxic substance
          response to carbohydrate
          response to inorganic substance
          negative regulation of peptidase activity
          negative regulation of collagen catabolic process
          negative regulation of extracellular matrix disassembly
          negative regulation of endopeptidase activity
          response to organic cyclic compound
          response to nutrient levels
          response to estradiol
          regulation of tissue remodeling
          cellular response to oxidative stress
          response to drug
          circadian sleep/wake cycle, REM sleep
          regulation of programmed cell death
          extracellular fibril organization
          positive regulation of DNA replication
          negative regulation of proteolysis
          response to axon injury
          Sertoli cell development
          negative regulation of elastin catabolic process
          negative regulation of blood vessel remodeling
          negative regulation of cell death
          cellular response to hydrogen peroxide
          beta-amyloid binding
          protease binding
          endopeptidase inhibitor activity
          cysteine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0