Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fosl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012954.1 | NP_037086.1 | 2 | 322 | ||
GenBank mRNA | AY622611.1 | - | 2 | 319 | ||
U18913.1 | - | 2 | 322 |