Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOG SIRNA Assays ›
          Gene Symbol:
          Nog
          Gene Name:
          noggin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 76811759 - 76813386 on Build Rnor_6.0
          UniGene ID:
          Rn.10154
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s129949

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nog
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012990.1 NP_037122.1 1 408
          GenBank mRNA BF404195.1 - 1 407

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_008711, BC132527, BC138148, AK146488.

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          osteoblast differentiation
          urogenital system development
          ureteric bud development
          in utero embryonic development
          endoderm formation
          mesoderm formation
          epithelial to mesenchymal transition
          neural plate morphogenesis
          neural tube closure
          pattern specification process
          axon guidance
          central nervous system development
          brain development
          motor neuron axon guidance
          dorsal/ventral pattern formation
          negative regulation of gene expression
          spinal cord development
          cell differentiation in hindbrain
          neural tube development
          pituitary gland development
          negative regulation of cell migration
          BMP signaling pathway
          regulation of BMP signaling pathway
          negative regulation of BMP signaling pathway
          forebrain development
          somatic stem cell population maintenance
          wound healing
          middle ear morphogenesis
          embryonic digit morphogenesis
          regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          axial mesoderm development
          notochord morphogenesis
          anatomical structure formation involved in morphogenesis
          embryonic skeletal system development
          negative regulation of astrocyte differentiation
          mesenchymal cell differentiation
          positive regulation of epithelial cell proliferation
          cartilage development
          negative regulation of cardiac muscle cell proliferation
          limb development
          embryonic skeletal joint morphogenesis
          negative regulation of cytokine activity
          face morphogenesis
          negative regulation of pathway-restricted SMAD protein phosphorylation
          lung morphogenesis
          prostatic bud formation
          ureteric bud formation
          fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
          negative regulation of cartilage development
          somite development
          cellular response to BMP stimulus
          negative regulation of canonical Wnt signaling pathway
          positive regulation of branching involved in ureteric bud morphogenesis
          positive regulation of glomerulus development
          regulation of fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
          negative regulation of apoptotic signaling pathway
          protein binding
          cytokine binding
          protein complex binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0