Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptprz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170685.1 | NP_001164156.1 | 12 | 3460 | ||
NM_013080.2 | NP_037212.2 | 12 | 3460 | |||
XM_006236137.2 | XP_006236199.1 | 12 | 3499 | |||
GenBank mRNA | U04998.1 | - | 12 | 3460 | ||
U09357.1 | - | 13 | 3460 |