Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTA1 SIRNA Assays ›
Gene Symbol:
Acta1
Gene Name:
actin, alpha 1, skeletal muscle
Gene Aliases:
-
Chromosome Location:
Chr.19: 56674072 - 56677084 on Build Rnor_6.0
UniGene ID:
Rn.82732
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s131419
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acta1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019212.2
NP_062085.1
5
697
GenBank mRNA
BC061974.1
-
5
697
5623909
FQ214583.1
-
5
706
FQ214585.1
-
5
708
FQ214598.1
-
5
706
FQ214600.1
-
5
706
FQ214603.1
-
5
706
FQ214610.1
-
3
251
FQ214623.1
-
5
706
FQ214676.1
-
5
706
FQ214680.1
-
5
706
FQ214697.1
-
5
706
FQ214743.1
-
5
706
FQ214756.1
-
5
706
FQ214779.1
-
5
706
FQ214807.1
-
5
706
FQ214814.1
-
5
706
FQ214822.1
-
5
706
FQ214831.1
-
5
706
FQ214832.1
-
5
706
FQ214839.1
-
5
709
FQ214841.1
-
5
706
FQ214842.1
-
5
706
FQ214857.1
-
5
710
FQ214859.1
-
5
706
FQ214860.1
-
5
706
FQ214866.1
-
5
706
FQ214883.1
-
5
705
FQ214904.1
-
5
706
FQ214905.1
-
5
705
FQ214916.1
-
5
706
FQ214921.1
-
5
706
FQ214936.1
-
5
706
FQ214948.1
-
5
706
FQ214949.1
-
5
706
FQ214957.1
-
5
706
FQ214969.1
-
5
706
FQ214975.1
-
5
708
FQ214976.1
-
5
706
FQ214978.1
-
5
706
FQ214990.1
-
5
706
FQ214993.1
-
5
706
FQ215012.1
-
5
706
FQ215025.1
-
5
706
FQ215038.1
-
5
706
FQ215058.1
-
5
706
FQ215089.1
-
5
706
FQ215153.1
-
5
706
FQ215158.1
-
5
704
FQ215173.1
-
5
706
FQ215204.1
-
5
704
FQ215212.1
-
5
706
FQ215215.1
-
5
706
FQ215241.1
-
5
706
FQ215249.1
-
5
707
FQ215253.1
-
5
706
FQ215256.1
-
5
706
FQ215275.1
-
5
706
FQ215280.1
-
5
706
FQ215282.1
-
5
706
FQ215298.1
-
5
708
FQ215324.1
-
5
706
FQ215333.1
-
5
706
FQ215342.1
-
5
706
FQ215346.1
-
5
706
FQ215366.1
-
5
706
FQ215380.1
-
5
706
FQ215381.1
-
5
706
FQ215393.1
-
5
706
FQ215396.1
-
5
706
FQ215398.1
-
5
706
FQ215402.1
-
5
706
FQ215403.1
-
5
706
FQ215405.1
-
5
706
FQ215414.1
-
5
708
FQ215424.1
-
5
706
FQ215431.1
-
5
706
FQ215433.1
-
5
705
FQ215434.1
-
5
708
FQ215438.1
-
5
706
FQ215446.1
-
5
706
FQ215449.1
-
5
708
FQ215454.1
-
5
706
FQ215464.1
-
5
706
FQ215465.1
-
5
706
FQ215468.1
-
5
706
FQ215470.1
-
5
706
FQ215474.1
-
5
706
FQ215483.1
-
5
706
FQ215488.1
-
5
706
FQ215518.1
-
5
715
FQ215523.1
-
5
706
FQ215554.1
-
5
706
FQ215572.1
-
5
706
FQ215593.1
-
5
706
FQ215628.1
-
5
706
FQ215631.1
-
3
320
FQ215655.1
-
5
704
FQ215673.1
-
5
706
FQ215674.1
-
5
706
FQ215690.1
-
5
706
FQ215785.1
-
5
706
FQ215799.1
-
5
706
FQ215809.1
-
5
706
FQ215813.1
-
5
706
FQ215835.1
-
5
706
FQ215836.1
-
5
706
FQ215839.1
-
5
706
FQ215851.1
-
5
706
FQ215852.1
-
5
704
FQ215864.1
-
5
707
FQ215888.1
-
5
705
FQ215893.1
-
5
707
FQ215896.1
-
5
706
FQ215907.1
-
5
708
FQ215924.1
-
3
398
FQ215950.1
-
5
706
FQ215964.1
-
5
706
FQ216016.1
-
5
706
FQ216035.1
-
5
706
FQ216043.1
-
5
706
FQ216050.1
-
5
706
FQ216139.1
-
5
706
FQ216168.1
-
5
704
FQ216196.1
-
5
706
FQ216224.1
-
5
706
FQ216246.1
-
5
706
FQ216276.1
-
5
706
FQ216278.1
-
5
708
FQ216287.1
-
5
704
FQ216291.1
-
5
706
FQ216293.1
-
5
706
FQ216305.1
-
5
706
FQ216329.1
-
5
706
FQ216333.1
-
5
706
FQ216350.1
-
5
706
FQ216352.1
-
5
706
FQ216360.1
-
5
706
FQ216369.1
-
5
708
FQ216403.1
-
5
706
FQ216509.1
-
5
704
FQ216586.1
-
5
706
FQ216603.1
-
5
706
FQ216606.1
-
5
706
FQ216607.1
-
5
702
FQ216610.1
-
5
704
FQ216620.1
-
5
706
FQ216635.1
-
5
704
FQ216639.1
-
5
706
FQ216644.1
-
5
706
FQ216664.1
-
5
704
FQ216675.1
-
5
706
FQ216676.1
-
5
706
FQ216692.1
-
5
706
FQ216703.1
-
5
706
FQ216713.1
-
5
706
FQ216721.1
-
5
706
FQ216723.1
-
5
706
FQ216725.1
-
5
702
FQ216739.1
-
5
706
FQ216762.1
-
5
704
FQ216767.1
-
5
706
FQ216776.1
-
5
706
FQ216777.1
-
5
706
FQ216779.1
-
5
708
FQ216793.1
-
5
706
FQ216799.1
-
5
704
FQ216801.1
-
5
706
FQ216814.1
-
5
700
FQ216819.1
-
5
704
FQ216916.1
-
5
706
FQ216918.1
-
3
349
FQ216961.1
-
5
706
FQ216994.1
-
5
706
FQ217073.1
-
5
709
FQ217091.1
-
5
705
FQ217114.1
-
5
706
FQ217122.1
-
5
706
FQ217167.1
-
5
706
FQ217173.1
-
5
706
FQ217188.1
-
5
706
FQ217273.1
-
5
705
FQ217280.1
-
5
826
FQ217327.1
-
5
706
FQ217329.1
-
5
706
FQ217347.1
-
5
706
FQ217353.1
-
5
706
FQ217367.1
-
5
706
FQ217397.1
-
5
706
FQ217441.1
-
5
706
FQ217443.1
-
5
708
FQ217475.1
-
5
706
FQ217525.1
-
5
706
FQ217535.1
-
5
706
FQ217572.1
-
5
706
FQ217593.1
-
5
706
FQ217629.1
-
5
706
FQ217641.1
-
5
703
FQ217762.1
-
5
706
FQ217826.1
-
5
706
FQ217884.1
-
5
704
FQ217905.1
-
5
706
FQ217908.1
-
5
706
FQ217919.1
-
5
706
FQ217937.1
-
5
706
FQ217969.1
-
5
708
FQ217990.1
-
5
706
FQ218005.1
-
2
106
FQ218019.1
-
5
706
FQ218034.1
-
5
706
FQ223563.1
-
5
706
FQ223681.1
-
5
708
FQ223686.1
-
5
706
FQ223702.1
-
5
706
FQ223778.1
-
3
220
FQ223898.1
-
5
706
FQ223914.1
-
5
706
FQ223929.1
-
5
706
FQ224009.1
-
5
706
FQ224082.1
-
5
706
FQ224141.1
-
5
707
FQ224167.1
-
5
706
FQ224359.1
-
5
706
FQ224377.1
-
5
706
FQ224419.1
-
5
706
FQ224552.1
-
5
706
FQ224691.1
-
5
704
FQ224777.1
-
5
706
FQ224840.1
-
5
704
FQ224842.1
-
5
706
FQ224862.1
-
5
706
FQ224869.1
-
5
706
FQ224886.1
-
5
706
FQ224890.1
-
5
704
FQ224902.1
-
5
706
FQ224929.1
-
5
704
FQ224936.1
-
5
706
FQ224942.1
-
5
708
FQ224946.1
-
3
234
FQ224948.1
-
5
708
FQ224962.1
-
5
706
FQ224971.1
-
5
706
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001272041, NM_009606, M12866, BC014877, X03766, and others.
Panther Classification:
Molecular Function -
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
response to mechanical stimulus
response to extracellular stimulus
response to lithium ion
cell growth
skeletal muscle thin filament assembly
skeletal muscle fiber adaptation
response to steroid hormone
skeletal muscle fiber development
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mkfsp:80/100.66.78.65:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0