Search
Search

SIGLEC1| Gene Symbol: SIGLEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_023068.3 | NP_075556.1 | 5 | 1030 | ||
| XM_006723610.3 | XP_006723673.1 | 6 | 1225 | |||
| XM_011529324.2 | XP_011527626.1 | 6 | 1222 | |||
| XM_011529325.2 | XP_011527627.1 | 6 | 1228 | |||
| XM_011529326.2 | XP_011527628.1 | 6 | 1224 | |||
| XM_011529327.2 | XP_011527629.1 | 5 | 1158 | |||
| XM_011529328.2 | XP_011527630.1 | 6 | 1170 | |||
| XM_011529329.1 | XP_011527631.1 | 5 | 2411 | |||
| XM_011529330.1 | XP_011527632.1 | 5 | 2411 | |||
| XM_011529331.1 | XP_011527633.1 | 5 | 2329 | |||
| XM_011529332.1 | XP_011527634.1 | 4 | 615 | |||
| XM_011529333.2 | XP_011527635.1 | 5 | 2411 | |||
| GenBank mRNA | AF230073.1 | - | 5 | 1030 | ||
| AY927519.1 | - | LIT3529 AY927519 |
4 | 938 | ||
| BC141884.1 | - | 5 | 1030 | |||