Search
Search

| Gene Symbol: Ugt2b37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770015.1 | XP_008768237.1 | 5 | 1309 | ||
| GenBank mRNA | FQ219518.1 | - | 5 | 683 | ||
| M33746.1 | - | 5 | 1287 | |||
| M33747.1 | - | 115 | ||||
| Gene Symbol: Ugt2b17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153314.2 | NP_695226.2 | 1276 | |||
| GenBank mRNA | M31109.1 | - | 1276 | |||
| Y00156.1 | - | 5 | 1320 | |||